CAS RN 39998-64-6
CAS RN 39998-64-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxymelatonin, TRC
CAS: 39998-64-6 Molecular Formula: C13H16N2O3 Molecular Weight (g/mol): 248.28 Synonym: N-[2-(4-hydroxy-5-methoxy-1H-indol-3-yl)ethyl]acetamide SMILES: CC(NCCC1=CNC2=C1C(O)=C(OC)C=C2)=O