missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzonitrile, 98+%

Product Code. p-7022852
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
Unit Size:
100g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384709 50 g 50g
11422468 100 g 100g
11394709 250 g 250g
11432468 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11384709 Supplier Thermo Scientific Alfa Aesar Supplier No. A15803.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 767-00-0 | C7H5NO | 119.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Hydroxybenzonitrile is used as an intermediate in organic synthesis. It is also used in cosmetic as an antiseptic and disinfectant. It is involved as a raw material for the preparation of bomoxynil and ioxynil.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 767-00-0
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00002312
InChI Key CVNOWLNNPYYEOH-UHFFFAOYSA-N
Synonym 4-cyanophenol, p-hydroxybenzonitrile, p-cyanophenol, benzonitrile, 4-hydroxy, benzonitrile, p-hydroxy, 4-cyano phenol, 4-hydroxybenzoic acid nitrile, 4-hydroxy-benzonitrile, 4-hydroxy benzonitrile, 4-hydroxybenzenecarbonitrile
PubChem CID 13019
ChEBI CHEBI:38622
IUPAC Name 4-hydroxybenzonitrile
SMILES C1=CC(=CC=C1C#N)O

Specifications

Melting Point 110°C to 114°C
Quantity 50 g
Beilstein 386130
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 119.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxybenzonitrile
compliance-icons
Product Identifier
  • 4-Cyanophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Cyanophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.