accessibility menu, dialog, popup

UserName

CAS RN 767-00-0

OHN

CAS RN 767-00-0

IUPAC Name: 4-hydroxybenzonitrile
Synonyms: 4-hydroxybenzonitrile P-cyanophenol 4-hydroxybenzoic acid nitrile
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: CVNOWLNNPYYEOH-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (6)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)