Learn More
4-Hydroxybenzenesulfonyl Chloride, TRC
Product Code. 30401500
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401500 250 mg 250mg
30391384 500 mg -
30397040 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30401500 Supplier Toronto Research Chemicals Supplier No. TRCH999963250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4025-67-6
Molecular Formula C6H5O3SCl
Molecular Weight (g/mol) 192.62
IUPAC Name 4-hydroxybenzenesulfonyl chloride
SMILES Oc1ccc(cc1)S(=O)(=O)Cl
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C6H5ClO3S/c7-11(9,10)6-3-1-5(8)2-4-6/h1-4,8H
Formula Weight 191.9648
Chemical Name or Material 4-Hydroxybenzenesulfonyl Chloride