CAS RN 4025-67-6
CAS RN 4025-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxybenzenesulfonyl Chloride, TRC
CAS: 4025-67-6 Molecular Formula: C6H5O3SCl Molecular Weight (g/mol): 192.62 IUPAC Name: 4-hydroxybenzenesulfonyl chloride SMILES: Oc1ccc(cc1)S(=O)(=O)Cl