Learn More
4-Hydroxy-1,3-phenylenediammonium Dichloride, Sigma-Aldrich™
Product Code. 15851801
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15851801 250 g 250g
15841801 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15851801 Supplier Sigma Aldrich Supplier No. 8223310250
 Restricted Item
Estimated Shipment: 24-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 137-09-7
Grade Research
Molecular Formula 4-(OH)C6H3-1,3-(NH2)2 · 2 HCl
Molecular Weight (g/mol) 197.06
Specifications
Melting Point 205°C
MDL Number MFCD00012979
Quantity 250 g
Synonym 4-Hydroxy-1,3-phenylenediammonium dichloride, Amidol, 2,4-Diaminophenol dihydrochloride, Phenol-2,4-diammonium dichloride, 1-Hydroxybenzene-2,4-diammonium dichloride, 4-Hydroxy-1,3-phenylenediammonium dichloride