accessibility menu, dialog, popup

UserName

CAS RN 137-09-7

HHClClNH2OHH2N

CAS RN 137-09-7

IUPAC Name: dihydrogen 2,4-diaminophenol dichloride
Synonyms: amidol
Molecular Weight (g/mol): 197.06
Molecular Formula: C6H10Cl2N2O
InChi Key: KQEIJFWAXDQUPR-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)