missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzenesulfonyl chloride, 97%

Product Code. p-7047615
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338886 1 g 1g
11348886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338886 Supplier Thermo Scientific Alfa Aesar Supplier No. L19517.03
Add to basket
This item is not returnable. View return policy

CAS: 7079-48-3 | C7H6ClFO2S | 208.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7079-48-3
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD03094390
InChI Key XLPGWKNCWMFHOD-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl
PubChem CID 2778602
IUPAC Name 4-fluoro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)F)S(=O)(=O)Cl

Specifications

Density 1.433
Boiling Point 245°C to 246°C
Flash Point >110°C (230°F)
Refractive Index 1.537
Quantity 1 g
UN Number UN3265
Beilstein 6720115
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.