accessibility menu, dialog, popup

UserName

CAS RN 7079-48-3

CH3FSOOCl

CAS RN 7079-48-3

IUPAC Name: 4-fluoro-2-methylbenzene-1-sulfonyl chloride
Synonyms: 4-fluoro-2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: XLPGWKNCWMFHOD-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(F)=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)