missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride, 99.5%, MP Biomedicals™

Product Code. 11476702 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476702 250 mg 250mg
11486702 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476702 Supplier MP Biomedicals Supplier No. 0215093183
  • ONLINE EXCLUSIVE PRICE
    77.15 EUR
    73.50 EUR / 250mg
    Save 3.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chromophoric label for amino acids, peptides, and proteins. Soluble in acetonitrile and DMF; slightly soluble in acetone

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56512-49-3
Molecular Formula C14H14ClN3O2S
Molecular Weight (g/mol) 323.80
MDL Number MFCD00007444
InChI Key VTVWTPGLLAELLI-UHFFFAOYSA-N
Synonym dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride
PubChem CID 91660
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonyl chloride
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Color Red
Quantity 250 mg
Formula Weight 323.8
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.