accessibility menu, dialog, popup

UserName

H3CNCH3NNSClOO(E/Z)

CAS RN 56512-49-3

IUPAC Name: 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonyl chloride
Synonyms: 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzenesulfonyl chloride
Molecular Weight (g/mol): 323.8
Molecular Formula: C14H14ClN3O2S
InChi Key: VTVWTPGLLAELLI-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)