missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)cinnamic acid, 97%, Thermo Scientific™

Product Code. 15494857
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494857 250 mg 250mg
15484857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494857 Supplier Thermo Scientific Alfa Aesar Supplier No. H32170.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127842-70-0
Molecular Formula C10H8F2O3
Molecular Weight (g/mol) 214.168
MDL Number MFCD01222822
InChI Key OWLKYWTYLFJOGX-ZZXKWVIFSA-N
Synonym 4-difluoromethoxy cinnamic acid, e-3-4-difluoromethoxy phenyl acrylic acid, 3-4-difluoromethoxy-phenyl-acrylic acid, 2e-3-4-difluoromethoxy phenyl prop-2-enoic acid, e-3-4-difluoromethoxy phenyl prop-2-enoic acid, 2-propenoic acid, 3-4-difluoromethoxy phenyl-, 2e
PubChem CID 703938
IUPAC Name (E)-3-[4-(difluoromethoxy)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)OC(F)F

Specifications

Melting Point 186°C to 189°C
Quantity 250 mg
Formula Weight 214.17
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)cinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.