accessibility menu, dialog, popup

UserName

CAS RN 127842-70-0

IUPAC Name: 3-[4-(difluoromethoxy)phenyl]prop-2-enoic acid
Synonyms: 3-[4-(difluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 214.17
Molecular Formula: C10H8F2O3
InChi Key: OWLKYWTYLFJOGX-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC=C(OC(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results