missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzamide, 97%

Product Code. p-7028339
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441627 1 g 1g
11451627 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441627 Supplier Thermo Scientific Alfa Aesar Supplier No. B25600.03
Add to basket
This item is not returnable. View return policy

CAS: 3034-34-2 | C8H6N2O | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-34-2
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00017133
InChI Key FUKWTMJZHKZKFA-UHFFFAOYSA-N
Synonym p-cyanobenzamide, benzamide, 4-cyano, unii-3p9ug7v78s, benzamide, p-cyano, benzamide, p-cyano-8ci, p-cyano benzamide, 4-cyano-benzamide, cyanobenzamide, acmc-1agnt, p-cyanobenzoic acid amide
PubChem CID 76427
IUPAC Name 4-cyanobenzamide
SMILES NC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 225°C to 228°C
Quantity 1 g
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.