CAS RN 3034-34-2
IUPAC Name:
4-cyanobenzamide
Synonyms:
P-cyanobenzamide
Molecular Weight (g/mol):
146.15
Molecular Formula:
C8H6N2O
InChi Key:
FUKWTMJZHKZKFA-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-Cyanobenzamide, 97%
CAS: 3034-34-2 Molecular Formula: C8H6N2O Molecular Weight (g/mol): 146.15 MDL Number: MFCD00017133 InChI Key: FUKWTMJZHKZKFA-UHFFFAOYSA-N Synonym: p-cyanobenzamide,benzamide, 4-cyano,unii-3p9ug7v78s,benzamide, p-cyano,benzamide, p-cyano-8ci,p-cyano benzamide,4-cyano-benzamide,cyanobenzamide,acmc-1agnt,p-cyanobenzoic acid amide PubChem CID: 76427 IUPAC Name: 4-cyanobenzamide SMILES: NC(=O)C1=CC=C(C=C1)C#N