missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-pentylbiphenyl, 99%

Product Code. p-7026066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444907 1 g 1g
11454907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444907 Supplier Thermo Scientific Alfa Aesar Supplier No. B21856.03
Add to basket
This item is not returnable. View return policy

CAS: 40817-08-1 | C18H19N | 249.357 g/mol

4-Cyano-4'-n-pentylbiphenyl is used in the preparation of 4'-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-4′-n-pentylbiphenyl is used in the preparation of 4′-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.

Solubility
Miscible with Acetone, chloroform, toluene, acetonitrile, methylene chloride. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40817-08-1
Molecular Formula C18H19N
Molecular Weight (g/mol) 249.357
MDL Number MFCD00036350
InChI Key HHPCNRKYVYWYAU-UHFFFAOYSA-N
Synonym 4-cyano-4'-pentylbiphenyl, 4'-pentyl-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-n-pentylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-pentyl, 4'-pentyl-4-biphenylcarbonitrile, 4-pentyl-4'-cyanobiphenyl, 4'-amyl-4-biphenylcarbonitrile, 4'-pentylbiphenyl-4-carbonitrile, 4-amyl-4'-cyanobiphenyl, 4-n-pentyl-4'-cyanobiphenyl
PubChem CID 92319
IUPAC Name 4-(4-pentylphenyl)benzonitrile
SMILES CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 22°C to 23°C
Density 1.008
Boiling Point 212°C to 215°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 1 g
UN Number UN3276
Solubility Information Miscible with Acetone,chloroform,toluene,acetonitrile,methylene chloride. Immiscible with water.
Formula Weight 249.36
Percent Purity 99%
Chemical Name or Material 4-Cyano-4'-n-pentylbiphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyano-4'-n-pentylbiphenyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Cyano-4'-n-pentylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.