accessibility menu, dialog, popup

UserName

H3CN

CAS RN 40817-08-1

IUPAC Name: 4'-pentyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4-cyano-4'-pentylbiphenyl
Molecular Weight (g/mol): 249.36
Molecular Formula: C18H19N
InChi Key: HHPCNRKYVYWYAU-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results