missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine, 96%

Product Code. 15428635
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428635 250 mg 250mg
15456627 1 g 1g
15466627 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428635 Supplier Thermo Scientific Alfa Aesar Supplier No. H33770.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 108831-68-1 | C8H7ClN2S | 198.67 g/mol

Employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108831-68-1
Molecular Formula C8H7ClN2S
Molecular Weight (g/mol) 198.67
MDL Number MFCD00464832
InChI Key HYOBKTVPACQCBR-UHFFFAOYSA-N
Synonym 4-chloro-5,6-dimethylthieno 2,3-d pyrimidine, 4-chloro-5,6-dimethyl-thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5,6-dimethyl, thieno 2,3-d pyrimidine, 4-chloro-5,6-dimethyl, 4-chloro-5,6-dimethylthiopheno 2,3-d pyrimidine, acmc-209vig, 4-chloranyl-5,6-dimethyl-thieno 2,3-d pyrimidine
PubChem CID 290212
IUPAC Name 4-chloro-5,6-dimethylthieno[2,3-d]pyrimidine
SMILES CC1=C(C)C2=C(Cl)N=CN=C2S1

Specifications

Melting Point 109°C to 110°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 198.68
Percent Purity 96%
Chemical Name or Material 4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.