CAS RN 108831-68-1
IUPAC Name:
4-chloro-5,6-dimethylthieno[2,3-d]pyrimidine
Synonyms:
4-chloro-5,6-dimethylthieno[2,3-d]pyrimidine
Molecular Weight (g/mol):
198.67
Molecular Formula:
C8H7ClN2S
InChi Key:
HYOBKTVPACQCBR-UHFFFAOYSA-N
SMILES:
CC1=C(C)C2=C(Cl)N=CN=C2S1
1
–
1
of
1
results
1
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine, 96%
CAS: 108831-68-1 Molecular Formula: C8H7ClN2S Molecular Weight (g/mol): 198.67 MDL Number: MFCD00464832 InChI Key: HYOBKTVPACQCBR-UHFFFAOYSA-N Synonym: 4-chloro-5,6-dimethylthieno 2,3-d pyrimidine,4-chloro-5,6-dimethyl-thieno 2,3-d pyrimidine,thieno 2,3-d pyrimidine,4-chloro-5,6-dimethyl,thieno 2,3-d pyrimidine, 4-chloro-5,6-dimethyl,4-chloro-5,6-dimethylthiopheno 2,3-d pyrimidine,acmc-209vig,4-chloranyl-5,6-dimethyl-thieno 2,3-d pyrimidine PubChem CID: 290212 IUPAC Name: 4-chloro-5,6-dimethylthieno[2,3-d]pyrimidine SMILES: CC1=C(C)C2=C(Cl)N=CN=C2S1