missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Product Code. 11438977
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438977 25 g 25g
11418977 1 g 1g
11428977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438977 Supplier Thermo Scientific Alfa Aesar Supplier No. B20580.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6294-93-5
Molecular Formula C7H4ClF3O
Molecular Weight (g/mol) 196.55
MDL Number MFCD00042525
InChI Key ZLFPIEUWXNRPNM-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenol, 2-chloro-5-hydroxybenzotrifluoride, phenol, 4-chloro-3-trifluoromethyl, 4-chloro-3-trifluoromethyl-phenol, phenol, p-chloro-m-trifluoromethyl, p-chloro-m-trifluoromethylphenol, 2-chloro-5-hydroxybezotrifluoride, pubchem2863
PubChem CID 80520
IUPAC Name 4-chloro-3-(trifluoromethyl)phenol
SMILES OC1=CC=C(Cl)C(=C1)C(F)(F)F

Specifications

Melting Point 42°C to 46°C
Boiling Point 120°C to 122°C (27 mmHg)
Odor Phenol-like
Quantity 25 g
UN Number UN3261
Beilstein 2261083
Formula Weight 196.56
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.