accessibility menu, dialog, popup

UserName

OHClFFF

CAS RN 6294-93-5

IUPAC Name: 4-chloro-3-(trifluoromethyl)phenol
Synonyms: 4-chloro-3-(trifluoromethyl)phenol
Molecular Weight (g/mol): 196.55
Molecular Formula: C7H4ClF3O
InChi Key: ZLFPIEUWXNRPNM-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C(=C1)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results