missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-chloro-5-methoxybenzeneboronic acid, 97%

Product Code. 15449827
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449827 1 g 1g
15420346 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449827 Supplier Alfa Aesar™ Supplier No. H53058.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 850567-94-1
Molecular Formula C7H7BBrClO3
Molecular Weight (g/mol) 265.29
MDL Number MFCD06659862
InChI Key DMUAPYZFKLNZMJ-UHFFFAOYSA-N
Synonym 4-bromo-2-chloro-5-methoxyphenyl boronic acid, 4-bromo-2-chloro-5-methoxy benzeneboronic acid, acmc-209q0w, b-2-chloro-4-bromo-5-methoxy-phenyl boronic acid, boronic acid,b-4-bromo-2-chloro-5-methoxyphenyl, b-4-bromo-2-chloro-5-methoxyphenyl-boronic acid, boronic acid, b-4-bromo-2-chloro-5-methoxyphenyl
PubChem CID 44118528
IUPAC Name (4-bromo-2-chloro-5-methoxyphenyl)boronic acid
SMILES COC1=C(Br)C=C(Cl)C(=C1)B(O)O

Specifications

Melting Point 260°C to 268°C
Quantity 1 g
Formula Weight 265.3
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-chloro-5-methoxybenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.