accessibility menu, dialog, popup

UserName

BClBrOCH3HOOH

CAS RN 850567-94-1

IUPAC Name: (4-bromo-2-chloro-5-methoxyphenyl)boronic acid
Synonyms: 4-bromo-2-chloro-5-methoxyphenylboronic acid
Molecular Weight (g/mol): 265.29
Molecular Formula: C7H7BBrClO3
InChi Key: DMUAPYZFKLNZMJ-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(Cl)C(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results