missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-(2-nitro-1-propenyl)thiophene, 97%

Product Code. 11496037
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496037 1 g 1g
11406047 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496037 Supplier Alfa Aesar™ Supplier No. B25060.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-19-3
Molecular Formula C7H6BrNO2S
Molecular Weight (g/mol) 248.094
MDL Number MFCD00052971
InChI Key DEFWRSIMNNRSKK-DJWKRKHSSA-N
Synonym 4-bromo-2-2-nitroprop-1-enyl thiophene, 4-bromo-2-2-nitro-1-propenyl thiophene, 4-bromo-2-z-2-nitroprop-1-enyl thiophene, 2-1z-2-nitroprop-1-enyl-4-bromothiophene
PubChem CID 5906915
IUPAC Name 4-bromo-2-[(Z)-2-nitroprop-1-enyl]thiophene
SMILES CC(=CC1=CC(=CS1)Br)[N+](=O)[O-]

Specifications

Melting Point 88°C to 90°C
Quantity 1 g
Formula Weight 248.09
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(2-nitro-1-propenyl)thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.