CAS RN 175205-19-3
IUPAC Name:
4-bromo-2-[(1E)-2-nitroprop-1-en-1-yl]thiophene
Synonyms:
4-bromo-2-[(1E)-2-nitroprop-1-en-1-yl]thiophene
Molecular Weight (g/mol):
248.09
Molecular Formula:
C7H6BrNO2S
InChi Key:
DEFWRSIMNNRSKK-GORDUTHDSA-N
SMILES:
C\C(=C/C1=CC(Br)=CS1)[N+]([O-])=O
1
–
1
of
1
results
1
Alfa Aesar™ 4-Bromo-2-(2-nitro-1-propenyl)thiophene, 97%
CAS: 175205-19-3 Molecular Formula: C7H6BrNO2S Molecular Weight (g/mol): 248.094 MDL Number: MFCD00052971 InChI Key: DEFWRSIMNNRSKK-DJWKRKHSSA-N Synonym: 4-bromo-2-2-nitroprop-1-enyl thiophene,4-bromo-2-2-nitro-1-propenyl thiophene,4-bromo-2-z-2-nitroprop-1-enyl thiophene,2-1z-2-nitroprop-1-enyl-4-bromothiophene PubChem CID: 5906915 IUPAC Name: 4-bromo-2-[(Z)-2-nitroprop-1-enyl]thiophene SMILES: CC(=CC1=CC(=CS1)Br)[N+](=O)[O-]