Learn More
4-Azido-L-phenylalanine Hydrochloride, TRC
Product Code. 17904495
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904495 25 mg 25mg
17971635 100 mg 100mg
17912945 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17904495 Supplier Toronto Research Chemicals Supplier No. TRCA92260025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34670-43-4
Molecular Formula C9 H10 N4 O2 . Cl H
Molecular Weight (g/mol) 242.662
Synonym 3-(p-Azidophenyl)-L-alanine Hydrochloride, 4-Azidophenylalanine Hydrochloride, p-Azido-L-phenylalanine Hydrochloride, p-Azidophenylalanine Hydrochloride
IUPAC Name (2S)-2-amino-3-(4-azidophenyl)propanoic acid;hydrochloride
SMILES Cl.N[C@@H](Cc1ccc(cc1)N=[N+]=[N-])C(=O)O
Specifications
InChI Formula InChI=1S/C9H10N4O2.ClH/c10-8(9(14)15)5-6-1-3-7(4-2-6)12-13-11;/h1-4,8H,5,10H2,(H,14,15);1H/t8-;/m0./s1
Quantity 25 mg
Formula Weight 242.0571
Shipping Condition Room Temperature
Chemical Name or Material 4-Azido-L-phenylalanine Hydrochloride