CAS RN 34670-43-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Azido-L-phenylalanine Hydrochloride, TRC
CAS: 34670-43-4 Molecular Formula: C9 H10 N4 O2 . Cl H Molecular Weight (g/mol): 242.662 Synonym: 3-(p-Azidophenyl)-L-alanine Hydrochloride,4-Azidophenylalanine Hydrochloride,p-Azido-L-phenylalanine Hydrochloride,p-Azidophenylalanine Hydrochloride IUPAC Name: (2S)-2-amino-3-(4-azidophenyl)propanoic acid;hydrochloride SMILES: Cl.N[C@@H](Cc1ccc(cc1)N=[N+]=[N-])C(=O)O
2
4-Azido-L-phenylalanine hydrochloride, MedChemExpress
MedChemExpress 4-Azido-L-phenylalanine hydrochloride is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments.