missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Azido-L-phenylalanine hydrochloride, MedChemExpress
Description
4-Azido-L-phenylalanine hydrochloride is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments.
Specifications
Specifications
| Chemical Name or Material | 4-Azido-L-phenylalanine hydrochloride |
| Physical Form | Solid |
| CAS | 34670-43-4 |
| Color | Khaki |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H11ClN4O2 |
| Synonym | p-Azidophenylalanine hydrochloride p-Azido-L-phenylalanine hydrochloride |
| Solubility Information | DMSO : ≥ 103.3 mg/mL (425.70 mM) ∣H2O : 10 mg/mL (41.21 mM; Need ultrasonic) |
| SMILES | O=C(O)[C@@H](N)CC1=CC=C(N=[N+]=[N-])C=C1.Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?