Learn More
4-Amino-3-chloro-5-methylbenzonitrile, tech. 90%, Thermo Scientific™
Product Code. 11363226
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363226 5 g 5g
11353226 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11363226 Supplier Thermo Scientific Alfa Aesar Supplier No. L12541.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 158296-69-6
Molecular Formula C8H7ClN2
Molecular Weight (g/mol) 166.608
MDL Number MFCD00041476
InChI Key NDTNVCCDQAOBSZ-UHFFFAOYSA-N
Synonym 4-amino-3-chloro-5-methyl-benzonitrile, benzonitrile, 4-amino-3-chloro-5-methyl, 4-amino-3-chloro-5-methylbenzenecarbonitrile, 2-chloro-4-cyano-6-methylaniline, acmc-1cuhq, ksc498c2h, 4-amino-3-chloro-5-methyl benzonitrile, 4-azanyl-3-chloranyl-5-methyl-benzenecarbonitrile
PubChem CID 2735301
IUPAC Name 4-amino-3-chloro-5-methylbenzonitrile
SMILES CC1=C(C(=CC(=C1)C#N)Cl)N
Specifications
Melting Point 128°C to 130°C
Quantity 5 g
UN Number UN3439
Beilstein 7527075
Formula Weight 166.61
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Amino-3-chloro-5-methylbenzonitrile