CAS RN 158296-69-6
IUPAC Name:
4-amino-3-chloro-5-methylbenzonitrile
Synonyms:
4-amino-3-chloro-5-methylbenzonitrile
Molecular Weight (g/mol):
166.61
Molecular Formula:
C8H7ClN2
InChi Key:
NDTNVCCDQAOBSZ-UHFFFAOYSA-N
SMILES:
CC1=C(N)C(Cl)=CC(=C1)C#N
1
–
1
of
1
results
1
4-Amino-3-chloro-5-methylbenzonitrile, tech. 90%, Thermo Scientific™
CAS: 158296-69-6 Molecular Formula: C8H7ClN2 Molecular Weight (g/mol): 166.608 MDL Number: MFCD00041476 InChI Key: NDTNVCCDQAOBSZ-UHFFFAOYSA-N Synonym: 4-amino-3-chloro-5-methyl-benzonitrile,benzonitrile, 4-amino-3-chloro-5-methyl,4-amino-3-chloro-5-methylbenzenecarbonitrile,2-chloro-4-cyano-6-methylaniline,acmc-1cuhq,ksc498c2h,4-amino-3-chloro-5-methyl benzonitrile,4-azanyl-3-chloranyl-5-methyl-benzenecarbonitrile PubChem CID: 2735301 IUPAC Name: 4-amino-3-chloro-5-methylbenzonitrile SMILES: CC1=C(C(=CC(=C1)C#N)Cl)N