missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloroveratrole, TRC

Product Code. 30388521
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388521 500 mg
30372182 100 mg
30379332 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388521 Supplier Toronto Research Chemicals Supplier No. TRCD437580500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2772-46-5
Molecular Formula C8H8O2Cl2
Molecular Weight (g/mol) 207.05
Synonym 4,5-Dichloroveratrole, Benzene, 1,2-dichloro-4,5-dimethoxy-, Veratrole, 4,5-dichloro- (6CI), 1,2-Dichloro-4,5-dimethoxybenzene, 4,5-Dichloro-1,2-dimethoxybenzene, 4,5-Dichloroveratrole
IUPAC Name 1,2-dichloro-4,5-dimethoxybenzene
SMILES COc1cc(Cl)c(Cl)cc1OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H8Cl2O2/c1-11-7-3-5(9)6(10)4-8(7)12-2/h3-4H,1-2H3
Formula Weight 205.9901
Chemical Name or Material 4,5-Dichloroveratrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.