CAS RN 2772-46-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4,5-Dichloroveratrole, TRC
CAS: 2772-46-5 Molecular Formula: C8H8O2Cl2 Molecular Weight (g/mol): 207.05 Synonym: 4,5-Dichloroveratrole,Benzene, 1,2-dichloro-4,5-dimethoxy-,Veratrole, 4,5-dichloro- (6CI),1,2-Dichloro-4,5-dimethoxybenzene,4,5-Dichloro-1,2-dimethoxybenzene,4,5-Dichloroveratrole IUPAC Name: 1,2-dichloro-4,5-dimethoxybenzene SMILES: COc1cc(Cl)c(Cl)cc1OC
2
4,5-Dichloroveratrole, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality