missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-methylisothiazol-3-one, TRC

Product Code. 30007493
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007493 10 mg 10mg
30004343 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007493 Supplier Toronto Research Chemicals Supplier No. TRCD43543010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26542-23-4
Molecular Formula C4 H3 Cl2 N O S
Molecular Weight (g/mol) 184.04
Synonym 3(2H)-Isothiazolone, 4,5-dichloro-2-methyl-, 4-Isothiazolin-3-one, 4,5-dichloro-2-methyl- (8CI), 4,5-Dichloro-2-methyl-3(2H)-isothiazolone, 2-Methyl-4,5-dichloro-3-isothiazolone, 2-Methyl-4,5-dichloroisothiazolin-3-one, 4,5-Dichloro-2-methyl-4-isothiazolin-3-one, 4,5-Dichloro-2-methylisothiazol-3-one
IUPAC Name 4,5-dichloro-2-methyl-1,2-thiazol-3-one
SMILES CN1SC(=C(Cl)C1=O)Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H3Cl2NOS/c1-7-4(8)2(5)3(6)9-7/h1H3
Formula Weight 182.9312
Percent Purity >95%
Chemical Name or Material 4,5-Dichloro-2-methylisothiazol-3-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.