CAS RN 26542-23-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4,5-Dichloro-2-methylisothiazol-3-one, TRC
CAS: 26542-23-4 Molecular Formula: C4 H3 Cl2 N O S Molecular Weight (g/mol): 184.04 Synonym: 3(2H)-Isothiazolone, 4,5-dichloro-2-methyl-,4-Isothiazolin-3-one, 4,5-dichloro-2-methyl- (8CI),4,5-Dichloro-2-methyl-3(2H)-isothiazolone,2-Methyl-4,5-dichloro-3-isothiazolone,2-Methyl-4,5-dichloroisothiazolin-3-one,4,5-Dichloro-2-methyl-4-isothiazolin-3-one,4,5-Dichloro-2-methylisothiazol-3-one IUPAC Name: 4,5-dichloro-2-methyl-1,2-thiazol-3-one SMILES: CN1SC(=C(Cl)C1=O)Cl
2
4,5-Dichloro-2-methylisothiazol-3-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.