missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-methylisothiazol-3-one, TRC

Product Code. 30004343
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004343 5 mg 5mg
30007493 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004343 Supplier Toronto Research Chemicals Supplier No. TRCD4354305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26542-23-4
Molecular Formula C4 H3 Cl2 N O S
Molecular Weight (g/mol) 184.04
Synonym 3(2H)-Isothiazolone, 4,5-dichloro-2-methyl-, 4-Isothiazolin-3-one, 4,5-dichloro-2-methyl- (8CI), 4,5-Dichloro-2-methyl-3(2H)-isothiazolone, 2-Methyl-4,5-dichloro-3-isothiazolone, 2-Methyl-4,5-dichloroisothiazolin-3-one, 4,5-Dichloro-2-methyl-4-isothiazolin-3-one, 4,5-Dichloro-2-methylisothiazol-3-one
IUPAC Name 4,5-dichloro-2-methyl-1,2-thiazol-3-one
SMILES CN1SC(=C(Cl)C1=O)Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C4H3Cl2NOS/c1-7-4(8)2(5)3(6)9-7/h1H3
Formula Weight 182.9312
Percent Purity >95%
Chemical Name or Material 4,5-Dichloro-2-methylisothiazol-3-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.