missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 15408605
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408605 5 g 5g
15418605 250 mg 250mg
15498595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408605 Supplier Thermo Scientific Alfa Aesar Supplier No. H33704.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863675-89-2
Molecular Formula C12H16F3N3
Molecular Weight (g/mol) 259.276
MDL Number MFCD09042650
InChI Key ILGSVUPRBCGGLF-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl-2-trifluoromethyl aniline, 4-4-methyl-1-piperazinyl-2-trifluoromethyl aniline
PubChem CID 16770363
IUPAC Name 4-(4-methylpiperazin-1-yl)-2-(trifluoromethyl)aniline
SMILES CN1CCN(CC1)C2=CC(=C(C=C2)N)C(F)(F)F

Specifications

Quantity 5 g
Sensitivity Air sensitive
Formula Weight 259.28
Percent Purity 97%
Chemical Name or Material 4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.