accessibility menu, dialog, popup

UserName

CH3NNNH2FFF

CAS RN 863675-89-2

IUPAC Name: 4-(4-methylpiperazin-1-yl)-2-(trifluoromethyl)aniline
Synonyms: 4-(4-methylpiperazin-1-yl)-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 259.28
Molecular Formula: C12H16F3N3
InChi Key: ILGSVUPRBCGGLF-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC(=C(N)C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results