Learn More
4-(4-Hydroxyphenyl)-2-methylthiazole, 97%, Thermo Scientific™
Product Code. 15494708
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. 15494708
Brand: Thermo Scientific Alfa Aesar H58411.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30686-73-8
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD02642878
InChI Key HOAYTTLLVORNLA-UHFFFAOYSA-N
Synonym 4-2-methyl-4-thiazolyl phenol, 4-2-methyl-1,3-thiazol-4-yl phenol, 4-2-methylthiazol-4-yl phenol, 4-4-hydroxyphenyl-2-methylthiazole, 4-2-methyl-4-thiazolyl phenol for biochemical research, 4-2-methyl-thiazol-4-yl-phenol, phenol, p-2-methyl-4-thiazolyl, 4-2-methyl-4-thiazolyl phenol*, 4-2-methyl-1,3-thiazol-4-yl phenol #, phenol, 4-2-methyl-4-thiazolyl-, hydrochloride
PubChem CID 591743
IUPAC Name 4-(2-methyl-1,3-thiazol-4-yl)phenol
SMILES CC1=NC(=CS1)C2=CC=C(C=C2)O
Specifications
Quantity 10 g
Beilstein 1104995
Sensitivity Air Sensitive
Formula Weight 191.25
Percent Purity 97%
Chemical Name or Material 4-(4-Hydroxyphenyl)-2-methylthiazole