CAS RN 30686-73-8
CAS RN 30686-73-8
IUPAC Name:
4-(2-methyl-1,3-thiazol-4-yl)phenol
Synonyms:
4-(2-methyl-1,3-thiazol-4-yl)phenol
Molecular Weight (g/mol):
191.25
Molecular Formula:
C10H9NOS
InChi Key:
HOAYTTLLVORNLA-UHFFFAOYSA-N
SMILES:
CC1=NC(=CS1)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
4-(4-Hydroxyphenyl)-2-methylthiazole, 97%, Thermo Scientific™
CAS: 30686-73-8 Molecular Formula: C10H9NOS Molecular Weight (g/mol): 191.248 MDL Number: MFCD02642878 InChI Key: HOAYTTLLVORNLA-UHFFFAOYSA-N Synonym: 4-2-methyl-4-thiazolyl phenol,4-2-methyl-1,3-thiazol-4-yl phenol,4-2-methylthiazol-4-yl phenol,4-4-hydroxyphenyl-2-methylthiazole,4-2-methyl-4-thiazolyl phenol for biochemical research,4-2-methyl-thiazol-4-yl-phenol,phenol, p-2-methyl-4-thiazolyl,4-2-methyl-4-thiazolyl phenol*,4-2-methyl-1,3-thiazol-4-yl phenol #,phenol, 4-2-methyl-4-thiazolyl-, hydrochloride PubChem CID: 591743 IUPAC Name: 4-(2-methyl-1,3-thiazol-4-yl)phenol SMILES: CC1=NC(=CS1)C2=CC=C(C=C2)O