missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-yl)benzoic acid, ≥90%, Thermo Scientific™

Product Code. p-2700436
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10384612 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10384612 Supplier Thermo Scientific Maybridge Supplier No. CC18501DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17616-04-5
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00586645
InChI Key LFIDZIWWYNTQOQ-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-yl benzoic acid, 4-imidazol-1-yl benzoic acid, 4-1-imidazolyl benzoic acid, benzoic acid, 4-1h-imidazol-1-yl, 4-1h-imidazole-1-yl benzoic acid, 4-imidazol-1-yl-benzoic acid, 4-imidazolylbenzoic acid, pubchem19564
PubChem CID 736529
IUPAC Name 4-imidazol-1-ylbenzoic acid
SMILES OC(=O)C1=CC=C(C=C1)N1C=CN=C1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 188.19
Percent Purity 90%
Chemical Name or Material 4-(1H-imidazol-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.