accessibility menu, dialog, popup

UserName

CAS RN 17616-04-5

IUPAC Name: 4-(1H-imidazol-1-yl)benzoic acid
Synonyms: 4-(imidazol-1-yl)benzoic acid
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: LFIDZIWWYNTQOQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results