Learn More
3-(Pentafluorothio)benzyl bromide, 97%, Thermo Scientific™
Product Code. 15466387
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466387 5 g 5g
15456387 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15466387 Supplier Thermo Scientific Alfa Aesar Supplier No. H33560.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1056893-24-3
Molecular Formula C7H6BrF5S
Molecular Weight (g/mol) 297.081
MDL Number MFCD16652411
InChI Key VBTCKIUDJZCRJU-UHFFFAOYSA-N
Synonym 3-pentafluorosulfur benzyl bromide, 1-bromomethyl-3-pentafluoro-??-sulfanyl benzene, 3-pentafluorothio benzyl bromide, 3-bromomethyl phenylpentafluorosulfur vi, 1-bromomethyl-3-pentafluoro-lambda∼6∼-sulfanyl benzene
PubChem CID 59393443
IUPAC Name [3-(bromomethyl)phenyl]-pentafluoro-$l^{6}-sulfane
SMILES C1=CC(=CC(=C1)S(F)(F)(F)(F)F)CBr
Specifications
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 297.08
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)benzyl bromide