accessibility menu, dialog, popup

UserName

SFFFFFBr

CAS RN 1056893-24-3

IUPAC Name: 1-(bromomethyl)-3-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-(bromomethyl)-3-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 297.08
Molecular Formula: C7H6BrF5S
InChi Key: VBTCKIUDJZCRJU-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC=CC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results