Learn More
3-Methylthiophene-2-carbonitrile, 96%
Product Code. 11420903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420903 25 g 25g
11400903 1 g 1g
11410903 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11420903 Supplier Thermo Scientific Alfa Aesar Supplier No. A13305.14
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
CAS: 55406-13-8 | C6H5NS | 123.173 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55406-13-8
Molecular Formula C6H5NS
Molecular Weight (g/mol) 123.173
MDL Number MFCD00052086
InChI Key ALZHYEITUZEZMT-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonitrile, 2-cyano-3-methylthiophene, acmc-1axot, 3-methyl-thiophene-2-carbonitrile, 3-methyl-2-thiophenecarbonitrile #
PubChem CID 521617
IUPAC Name 3-methylthiophene-2-carbonitrile
SMILES CC1=C(SC=C1)C#N
Specifications
Boiling Point 91°C to 93°C (12 mmHg)
Refractive Index 1.553
Quantity 25 g
UN Number UN3276
Beilstein 5495606
Formula Weight 123.18
Percent Purity 96%
Chemical Name or Material 3-Methylthiophene-2-carbonitrile