CAS RN 55406-13-8
IUPAC Name:
3-methylthiophene-2-carbonitrile
Synonyms:
3-methylthiophene-2-carbonitrile
Molecular Weight (g/mol):
123.17
Molecular Formula:
C6H5NS
InChi Key:
ALZHYEITUZEZMT-UHFFFAOYSA-N
SMILES:
CC1=C(SC=C1)C#N
1
–
1
of
1
results
1
3-Methylthiophene-2-carbonitrile, 96%
CAS: 55406-13-8 Molecular Formula: C6H5NS Molecular Weight (g/mol): 123.173 MDL Number: MFCD00052086 InChI Key: ALZHYEITUZEZMT-UHFFFAOYSA-N Synonym: 3-methyl-2-thiophenecarbonitrile,2-cyano-3-methylthiophene,acmc-1axot,3-methyl-thiophene-2-carbonitrile,3-methyl-2-thiophenecarbonitrile # PubChem CID: 521617 IUPAC Name: 3-methylthiophene-2-carbonitrile SMILES: CC1=C(SC=C1)C#N