Learn More
3-Methylpyridine-2-carboxaldehyde, 96%
Product Code. 15412608
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412608 1 g 1g
15422608 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15412608 Supplier Thermo Scientific Alfa Aesar Supplier No. H56168.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
CAS: 55589-47-4 | C7H7NO | 121.139 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55589-47-4
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD02181144
InChI Key JDYVLWWFVYNMTN-UHFFFAOYSA-N
Synonym 3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde
PubChem CID 1501874
IUPAC Name 3-methylpyridine-2-carbaldehyde
SMILES CC1=C(N=CC=C1)C=O
Specifications
Density 1.08
Flash Point 73°C (163°F)
Refractive Index 1.538
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 121.14
Percent Purity 97%
Chemical Name or Material 3-Methylpyridine-2-carboxaldehyde