accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 55589-47-4

IUPAC Name: 3-methylpyridine-2-carbaldehyde
Synonyms: 3-methylpyridine-2-carbaldehyde
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: JDYVLWWFVYNMTN-UHFFFAOYSA-N
SMILES: CC1=C(C=O)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results