Learn More
3-Methylpyridine-2-carboxaldehyde, 97%, Thermo Scientific™
Product Code. 10389407 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10389407 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10389407 Supplier Thermo Scientific Acros Supplier No. 429920010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 55589-47-4
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0800g/mL
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H7NO
MDL Number MFCD02181144
Quantity 1 g
Synonym 3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde
InChI Key JDYVLWWFVYNMTN-UHFFFAOYSA-N
SMILES CC1=C(N=CC=C1)C=O
IUPAC Name 3-methylpyridine-2-carbaldehyde
Molecular Weight (g/mol) 121.139
PubChem CID 1501874
Formula Weight 121.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methylpyridine-2-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylpyridine-2-carboxaldehyde, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.