Learn More
3-Methylphosphinicopropionic Acid Disodium Salt, TRC
Product Code. 30009143
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009143 100 mg 100mg
17944565 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009143 Supplier Toronto Research Chemicals Supplier No. TRCM326160100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66992-42-5
Molecular Formula C4H7Na2O4P
Molecular Weight (g/mol) 196.05
Synonym 3-(Hydroxymethylphosphinyl)propanoic Acid Disodium Salt, (2-Carboxyethyl)methylphosphinic Acid Disodium Salt, MPP Disodium Salt, MPPA Disodium Salt, Methylphosphinicopropionic Acid Disodium Salt;
IUPAC Name disodium;3-[methyl(oxido)phosphoryl]propanoate
SMILES O=P(CCC(O[Na])=O)(O[Na])C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C4H9O4P.2Na/c1-9(7,8)3-2-4(5)6;;/h2-3H2,1H3,(H,5,6)(H,7,8);;/q;2*+1/p-2
Formula Weight 195.9877
Percent Purity >95%
Chemical Name or Material 3-Methylphosphinicopropionic Acid Disodium Salt
Grade HPLC