CAS RN 66992-42-5
CAS RN 66992-42-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methylphosphinicopropionic Acid Disodium Salt, TRC
CAS: 66992-42-5 Molecular Formula: C4H7Na2O4P Molecular Weight (g/mol): 196.05 Synonym: 3-(Hydroxymethylphosphinyl)propanoic Acid Disodium Salt,(2-Carboxyethyl)methylphosphinic Acid Disodium Salt,MPP Disodium Salt,MPPA Disodium Salt,Methylphosphinicopropionic Acid Disodium Salt; IUPAC Name: disodium;3-[methyl(oxido)phosphoryl]propanoate SMILES: O=P(CCC(O[Na])=O)(O[Na])C